Learn More
4'-Methoxybiphenyl-4-sulfonyl chloride, 95%, Thermo Scientific™
Catalog Number p-7077341
Click to view available options
Quantity:
1 g
5 g
202752-04-3
C13H11ClO3S
282.74
MFCD01631917
AJWXJCPSCXFPJJ-UHFFFAOYSA-N
4'-methoxybiphenyl-4-sulfonyl chloride, 4'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonyl chloride, 4'-methoxy-biphenyl-4-sulfonyl chloride, 4'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl phenyl sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-methoxy, 4'-methoxy biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonylchloride, 4-4-methoxyphenyl phenyl sulfonylchloride
2794750
4'-methoxy-[1,1'-biphenyl]-4-sulfonyl chloride
COC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
202752-04-3
C13H11ClO3S
282.74
MFCD01631917
AJWXJCPSCXFPJJ-UHFFFAOYSA-N
4'-methoxybiphenyl-4-sulfonyl chloride, 4'-methoxy 1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonyl chloride, 4'-methoxy-biphenyl-4-sulfonyl chloride, 4'-methoxy-1,1'-biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl phenyl sulfonyl chloride, 1,1'-biphenyl-4-sulfonylchloride, 4'-methoxy, 4'-methoxy biphenyl-4-sulfonyl chloride, 4-4-methoxyphenyl benzenesulfonylchloride, 4-4-methoxyphenyl phenyl sulfonylchloride
2794750
4'-methoxy-[1,1'-biphenyl]-4-sulfonyl chloride
COC1=CC=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
Melting Point 104.0°C to 109.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 282.75
Percent Purity 95%
Chemical Name or Material 4'-Methoxybiphenyl-4-sulfonyl chloride
compliance-icons
Product Identifier
  • 4'-Methoxybiphenyl-4-sulfonyl chloride, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.