Learn More
4-Methoxybenzyl isothiocyanate, 94%, Thermo Scientific Chemicals
Catalog Number p-7045880
Click to view available options
Quantity:
1 g
5 g
3694-57-3
C9H9NOS
179.237
MFCD00041117
IMFQYAJJXFXVMM-UHFFFAOYSA-N
4-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxybenzene, 4-methoxybenzylisothiocyanate, benzene, 1-isothiocyanatomethyl-4-methoxy, p-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxy-benzene, chembl81201, 4-methoxybenzyl isothiocyanate 5gr, acmc-1cq6l
123197
1-(isothiocyanatomethyl)-4-methoxybenzene
COC1=CC=C(C=C1)CN=C=S
3694-57-3
C9H9NOS
179.237
MFCD00041117
IMFQYAJJXFXVMM-UHFFFAOYSA-N
4-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxybenzene, 4-methoxybenzylisothiocyanate, benzene, 1-isothiocyanatomethyl-4-methoxy, p-methoxybenzyl isothiocyanate, 1-isothiocyanatomethyl-4-methoxy-benzene, chembl81201, 4-methoxybenzyl isothiocyanate 5gr, acmc-1cq6l
123197
1-(isothiocyanatomethyl)-4-methoxybenzene
COC1=CC=C(C=C1)CN=C=S
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Methoxybenzyl isothiocyanate is used as pharmaceutical intermediates.
Solubility
Hydrolyzes in water.
Notes
Moisture Sensitive. Incompatible with bases, alcohols and amines. Store under dry inert gas. Protect from humidity and water. Store away from moisture.
TRUSTED_SUSTAINABILITY
Density 1.15
Boiling Point 132°C to 133°C (0.7 mmHg)
Odor Irritating
Refractive Index 1.5975
Quantity 1 g
UN Number UN2922
Beilstein 2089549
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 179.24
Percent Purity 94%
Chemical Name or Material 4-Methoxybenzyl isothiocyanate