Learn More
4-Isobutylstyrene, Mikromol™
Product Code. 17933496
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. 17933496
Brand: Mikromol MM0002.48
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
4-Isobutylstyrene
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-Isobutylstyrene
CAS 63444-56-4
Analyte or Component Names 4-Isobutylstyrene
Shipping Condition Ice pack (-20°C)
Molecular Formula C12 H16
Certifications/Compliance ISO 9001
Synonym 4-Isobutylstyrene
InChI Formula InChI=1S/C12H16/c1-4-11-5-7-12(8-6-11)9-10(2)3/h4-8,10H,1,9H2,2-3H3
SMILES CC(C)Cc1ccc(C=C)cc1
IUPAC Name 1-ethenyl-4-(2-methylpropyl)benzene
Molecular Weight (g/mol) 160.26
Quantity 10 mg
Formula Weight 160.1252 g/mol