Learn More
4-Iodoopyridin-2(1H)-one, 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97+% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | 4-iodo-1,2-dihydropyridin-2-one |
Formula Weight | 220.99 |
Percent Purity | 97% |
Chemical Name or Material | 4-iodo-2-pyridone |