missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-m-xylene, 98%, Thermo Scientific Chemicals
362.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.
Solubility
Not miscible or difficult to mix in water.
Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
4-Iodo-m-xylene is a reactant used in the preparation of paralog-selective Hsp90 inhibitors.
Solubility
Not miscible or difficult to mix in water.
Notes
Light Sensitive. Protect from light. Store away from oxidizing agents.
Specifications
Density | 1.628 |
Boiling Point | 231°C to 232°C |
Quantity | 10 g |
Refractive Index | 1.598 |
Beilstein | 1422602 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 232.07 |
Percent Purity | 98% |
Chemical Name or Material | 4-Iodo-m-xylene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'