missing translation for 'onlineSavingsMsg'
Learn More

(4-Imidazolyl)acetonitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-7031797
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
18502-05-1
C5H5N3
107.116
MFCD00070306
DQZBHUXBFNBJLX-UHFFFAOYSA-N
2-1h-imidazol-4-yl acetonitrile, 1h-imidazole-5-acetonitrile, 1h-imidazole-4-acetonitrile, 2-1h-imidazol-5-yl acetonitrile, 1h-imidazol-4-yl-acetonitrile, 4 5-cyanomethylimidazole, 1h-imidazol-4-yl acetonitrile, 2-3h-imidazol-4-yl acetonitrile, 4-imidazolyl acetonitrile, 1h imidazole-4-acetonitrile
571973
2-(1H-imidazol-5-yl)acetonitrile
C1=C(NC=N1)CC#N
This item is not returnable. View return policy
18502-05-1
C5H5N3
107.116
MFCD00070306
DQZBHUXBFNBJLX-UHFFFAOYSA-N
2-1h-imidazol-4-yl acetonitrile, 1h-imidazole-5-acetonitrile, 1h-imidazole-4-acetonitrile, 2-1h-imidazol-5-yl acetonitrile, 1h-imidazol-4-yl-acetonitrile, 4 5-cyanomethylimidazole, 1h-imidazol-4-yl acetonitrile, 2-3h-imidazol-4-yl acetonitrile, 4-imidazolyl acetonitrile, 1h imidazole-4-acetonitrile
571973
2-(1H-imidazol-5-yl)acetonitrile
C1=C(NC=N1)CC#N

(4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(4-Imidazolyl)acetonitrile is used to produce 2-(1H-imidazol-4-yl)-3-phenyl-acrylonitrile at ambient temperature.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 136°C to 138°C
Odor Odorless
Quantity 1 g
UN Number UN3439
Solubility Information Insoluble in water.
Formula Weight 107.11
Percent Purity 97%
Chemical Name or Material (4-Imidazolyl)acetonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'