Learn More
4-Hydroxybenzenesulfonic acid, 65%, Thermo Scientific Chemicals
Catalog Number p-7053432
Click to view available options
Quantity:
250 mL
1 L
98-67-9
C6H6O4S
174.17
MFCD00007506
FEPBITJSIHRMRT-UHFFFAOYSA-N
p-phenolsulfonic acid, phenolsulfonic acid, 4-phenolsulfonic acid, p-sulfophenol, sulfocarbolic acid, benzenesulfonic acid, 4-hydroxy, p-hydroxybenzenesulfonic acid, phenol-4-sulfonic acid, benzenesulfonic acid, p-hydroxy, 4-hydroxyphenylsulfonic acid
4765
CHEBI:32354
4-hydroxybenzenesulfonic acid
OC1=CC=C(C=C1)S(O)(=O)=O
98-67-9
C6H6O4S
174.17
MFCD00007506
FEPBITJSIHRMRT-UHFFFAOYSA-N
p-phenolsulfonic acid, phenolsulfonic acid, 4-phenolsulfonic acid, p-sulfophenol, sulfocarbolic acid, benzenesulfonic acid, 4-hydroxy, p-hydroxybenzenesulfonic acid, phenol-4-sulfonic acid, benzenesulfonic acid, p-hydroxy, 4-hydroxyphenylsulfonic acid
4765
CHEBI:32354
4-hydroxybenzenesulfonic acid
OC1=CC=C(C=C1)S(O)(=O)=O
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Density 1.337 g/mL
Refractive Index 1.489
Quantity 250 mL
UN Number UN1803
Beilstein 1869034
Solubility Information Soluble in water.
Formula Weight 174.17
Percent Purity 65%
Physical Form Liquid
Chemical Name or Material 4-Hydroxybenzenesulfonic acid