missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrophenylacetic acid, 99%, Thermo Scientific™

Catalog Number p-146637
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
10463-20-4
C8H7NO5
197.15
QBHBHOSRLDPIHG-UHFFFAOYSA-N
4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
447364
CHEBI:546274
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O
This item is not returnable. View return policy
10463-20-4
C8H7NO5
197.15
QBHBHOSRLDPIHG-UHFFFAOYSA-N
4-hydroxy-3-nitrophenylacetic acid, 2-4-hydroxy-3-nitrophenyl acetic acid, 4-hydroxy-3-nitrophenyl acetic acid, 3-nitro-4-hydroxyphenylacetic acid, 3-nitro-4-hydroxy phenylacetic acid, benzeneacetic acid, 4-hydroxy-3-nitro, no2hpa, hydroxynitrophenylaceticacid, acmc-2098cj, epitope id:158629
447364
CHEBI:546274
C1=CC(=C(C=C1CC(=O)O)[N+](=O)[O-])O
CAS Min % 98.5
CAS Max % 100.0
Melting Point 144.0°C to 147.0°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H3(NO2)CH2CO2H
Quantity 1 g
Beilstein 10,IV,562
Formula Weight 197.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Hydroxy-3-nitrophenylacetic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Hydroxy-3-nitrophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'