Learn More
4-Heptanone, 98%, Thermo Scientific Chemicals
Catalog Number p-140229
Click to view available options
Quantity:
5 mL
500 mL
2.5 L
Packaging:
Glass bottle
123-19-3
C7H14O
114.19
MFCD00009403
HCFAJYNVAYBARA-UHFFFAOYSA-N
4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
31246
heptan-4-one
CCCC(=O)CCC
123-19-3
C7H14O
114.19
MFCD00009403
HCFAJYNVAYBARA-UHFFFAOYSA-N
4-heptanone, dipropyl ketone, propyl ketone, butyrone, di-n-propyl ketone, 4-heptanone natural, 4-oxoheptane, unii-9bn582jq61, fema no. 2546, dipropylketon
31246
heptan-4-one
CCCC(=O)CCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -33.0°C
Color Colorless to Yellow
Density 0.8100g/mL
Boiling Point 145.0°C
Flash Point 48°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2COCH2CH2CH3
Refractive Index 1.4060 to 1.4080
Quantity 5 mL
Beilstein 01,699
Merck Index 15,3386
Specific Gravity 0.81
Solubility Information Solubility in water: 4.6g/L (20°C). Other solubilities: soluble in alcohol and ether
Viscosity 0.73 mPa.s (20°C)
Formula Weight 114.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Heptanone, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 4-Heptanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
Hazard Statement
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.