missing translation for 'onlineSavingsMsg'
Learn More

4'-Fluorobiphenyl-4-carboxylic acid, 96%, Thermo Scientific Chemicals

Catalog Number p-7031934
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
5731-10-2
C13H8FO2
215.20
MFCD01631909
LXWNTLBMNCXRQN-UHFFFAOYSA-M
4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
2782689
4-(4-fluorophenyl)benzoic acid
C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O
This item is not returnable. View return policy
5731-10-2
C13H8FO2
215.20
MFCD01631909
LXWNTLBMNCXRQN-UHFFFAOYSA-M
4-4-fluorophenyl benzoic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4'-fluorobiphenyl-4-carboxylic acid, 4'-fluoro-1,1'-biphenyl-4-carboxylic acid, 4-biphenyl-4'-fluoro-carboxylic acid, 4'-fluoro-biphenyl-4-carboxylic acid, 4-fluoro-biphenyl-4-carboxylic acid, 4'-fluoro 1,1'-biphenyl-4-carboxylic acid, 4fc
2782689
4-(4-fluorophenyl)benzoic acid
C1=CC(=CC=C1C2=CC=C(C=C2)F)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 243°C to 244°C
Quantity 1 g
Formula Weight 216.21
Percent Purity 96%
Chemical Name or Material 4'-Fluorobiphenyl-4-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'