missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzotrifluoride, 98+%, Thermo Scientific Chemicals
513.62 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
4-Fluorobenzotrifluoride was used in the synthesis of 2,2?-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2?-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluorobenzotrifluoride was used in the synthesis of 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
4-Fluorobenzotrifluoride was used in the synthesis of 2,2′-bis(trifluoromethylphenoxy)biphenyl via nucleophilic aromatic substitution reaction with 2,2′-biphenol, (S)-fluoxetine, potent inhibitor of neuronal serotonin-uptake and 1-aryloxy-2-substituted aminomethyltetrahydronaphthalene derivatives.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Melting Point | -42°C |
Density | 1.295 |
Boiling Point | 103°C to 105°C |
Flash Point | 10°C (50°F) |
Refractive Index | 1.401 |
Quantity | 5 g |
UN Number | UN1993 |
Beilstein | 1909390 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 164.1 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'