Learn More
4-Fluoro-2-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
Catalog Number p-7024834
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Questo articolo non è restituibile. Consulta la politica di reso
189807-21-4
C8H3ClF4O
226.55
MFCD00061158
OEYHURRIOWWRMD-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-chlorocarbonyl-5-fluorobenzotrifluoride, benzoyl chloride, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, acmc-1bwn8, ksc540e2h, attercop-chm at107069, 2-trifluoromethyl-4-fluorobenzoylchlorid
605680
4-fluoro-2-(trifluoromethyl)benzoyl chloride
FC1=CC=C(C(Cl)=O)C(=C1)C(F)(F)F
Questo articolo non è restituibile. Consulta la politica di reso
189807-21-4
C8H3ClF4O
226.55
MFCD00061158
OEYHURRIOWWRMD-UHFFFAOYSA-N
4-fluoro-2-trifluoromethyl benzoyl chloride, 4-fluoro-2-trifluoromethyl benzoylchloride, 2-trifluoromethyl-4-fluorobenzoyl chloride, 2-chlorocarbonyl-5-fluorobenzotrifluoride, benzoyl chloride, 4-fluoro-2-trifluoromethyl, alpha,alpha,alpha,4-tetrafluoro-o-toluoyl chloride, acmc-1bwn8, ksc540e2h, attercop-chm at107069, 2-trifluoromethyl-4-fluorobenzoylchlorid
605680
4-fluoro-2-(trifluoromethyl)benzoyl chloride
FC1=CC=C(C(Cl)=O)C(=C1)C(F)(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Fluoro-2-(trifluoromethyl)benzoyl chloride is used in medicine.
Solubility
Not miscible in water.
Notes
Store at 4°C. Store away from moisture and oxidizing agents under dry inert gas. Incompatible with active metals, bases and amines. Moisture Sensitive.
TRUSTED_SUSTAINABILITY
Density 1.495
Boiling Point 157°C
Flash Point 85°C (185°F)
Refractive Index 1.468
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Not miscible in water.
Formula Weight 226.56
Percent Purity 97%
Chemical Name or Material 4-Fluoro-2-(trifluoromethyl)benzoyl chloride
Vedi altri risultati Mostra meno risultati