missing translation for 'onlineSavingsMsg'
Learn More

4-Ethylmorpholine, 98%, Thermo Scientific Chemicals

Catalog Number p-7081702
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1000 mL
This item is not returnable. View return policy
100-74-3
C6H13NO
MFCD00006177
HVCNXQOWACZAFN-UHFFFAOYSA-N
7525
4-ethylmorpholine
CCN1CCOCC1
This item is not returnable. View return policy
100-74-3
C6H13NO
MFCD00006177
HVCNXQOWACZAFN-UHFFFAOYSA-N
7525
4-ethylmorpholine
CCN1CCOCC1

N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Ethylmorpholine is a component of the buffer used in basic peptide separation through anion-exchange chromatography. Further, it acts as a catalyst in the preparation of polyurethane foam.

Solubility
Miscible with water.

Notes
Light and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -63°C
Density 0.908
Boiling Point 138°C to 139°C
Flash Point 27°C
Refractive Index 1.4413
Quantity 100 mL
UN Number UN2734
Beilstein 102969
Sensitivity Air Sensitive
Synonym n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq
Solubility Information Miscible with water.
Molecular Weight (g/mol) 115.18
Formula Weight 115.18
Percent Purity 98%
Chemical Name or Material 4-Ethylmorpholine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'