Learn More
Alfa Aesar™ 4-Ethylbenzonitrile, 98%
Brand: Alfa Aesar™ L07921.18
Additional Details : CAS Number : 25309-65-3 Weight : 0.05000kg
Quantity | 50g |
---|
Chemical Identifiers
25309-65-3 | |
131.178 | |
SXFFMFAQNAFSLF-UHFFFAOYSA-N | |
91393 | |
CCC1=CC=C(C=C1)C#N |
C9H9N | |
MFCD00016383 | |
benzonitrile, 4-ethyl, p-ethylbenzonitrile, 1-cyano-4-ethylbenzene, 4-ethylbenzenecarbonitrile, 4-ethyl-benzonitrile, acmc-20amj0, 4-ethylbenzonitrile | |
4-ethylbenzonitrile |
Specifications
4-Ethylbenzonitrile | |
97°C (206°F) | |
237°C | |
25309-65-3 | |
MFCD00016383 | |
benzonitrile, 4-ethyl, p-ethylbenzonitrile, 1-cyano-4-ethylbenzene, 4-ethylbenzenecarbonitrile, 4-ethyl-benzonitrile, acmc-20amj0, 4-ethylbenzonitrile | |
CCC1=CC=C(C=C1)C#N | |
131.178 | |
131.18 |
0.956 | |
1.526 | |
50g | |
C9H9N | |
2040912 | |
SXFFMFAQNAFSLF-UHFFFAOYSA-N | |
4-ethylbenzonitrile | |
91393 | |
98% |