missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,3-difluorobenzaldehyde, 97%, Thermo Scientific™

Catalog Number p-7031385
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
126162-95-6
C9H8F2O2
186.16
MFCD09258700
DYASUFNACNHJMK-UHFFFAOYSA-N
2,3-difluoro-4-ethoxybenzaldehyde, 4-ethoxy-2,3-difluoro-benzaldehyde, 4-ethoxy-2,3-difluoro benzaldehyde
14495625
4-ethoxy-2,3-difluorobenzaldehyde
CCOC1=C(F)C(F)=C(C=O)C=C1
This item is not returnable. View return policy
126162-95-6
C9H8F2O2
186.16
MFCD09258700
DYASUFNACNHJMK-UHFFFAOYSA-N
2,3-difluoro-4-ethoxybenzaldehyde, 4-ethoxy-2,3-difluoro-benzaldehyde, 4-ethoxy-2,3-difluoro benzaldehyde
14495625
4-ethoxy-2,3-difluorobenzaldehyde
CCOC1=C(F)C(F)=C(C=O)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Ethoxy-2,3-difluorobenzaldehyde, is used as a pharmaceutical intermediate in antibiotics, as sedatives, and for cancer treatment.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 70°C to 73°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 186.16
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'