missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Cyanobenzoic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.
Solubility
Soluble in methanol.
Notes
Air Sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases.
4-Cyanobenzoic acid is used as an inhibitor. 4-cyanobenzaldehyde and 4-cyanobenzoic acid can inhibit both the monophenolase activity and the diphenolase activity of mushroom tyrosinase.
Solubility
Soluble in methanol.
Notes
Air Sensitive. Incompatible with strong oxidizing agents, strong acids, strong bases.
Specifications
Melting Point | ∼221°C (decomposition) |
Quantity | 5 g |
Beilstein | 1862865 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in methanol. |
Formula Weight | 147.13 |
Percent Purity | 98% |
Chemical Name or Material | 4-Cyanobenzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'