missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorothiobenzamide, 97%, Thermo Scientific Chemicals

Catalog Number p-7081852
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
This item is not returnable. View return policy
2521-24-6
C7H6ClNS
171.64
MFCD00040956
OKPUICCJRDBRJT-UHFFFAOYSA-N
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
2734826
NC(=S)C1=CC=C(Cl)C=C1
This item is not returnable. View return policy
2521-24-6
C7H6ClNS
171.64
MFCD00040956
OKPUICCJRDBRJT-UHFFFAOYSA-N
4-chlorothiobenzamide, 4-chlorobenzothioamide, p-chlorothiobenzamide, p-chlorobenzothiamide, 4-chlorobenzene-1-carbothioamide, 4-chloro-thiobenzamide, benzenecarbothioamide, 4-chloro, benzamide, p-chlorothio, benzamide, p-chlorothio-8ci, amino 4-chlorophenyl methane-1-thione
2734826
NC(=S)C1=CC=C(Cl)C=C1

4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorothiobenzamide is used as a catalytic agent and as petrochemical additive.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 123°C to 129°C
Quantity 10 g
Beilstein 606603
Solubility Information Insoluble in water.
IUPAC Name 4-chlorobenzenecarbothioamide
Formula Weight 171.65
Percent Purity 97%
Chemical Name or Material 4-Chlorothiobenzamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'