missing translation for 'onlineSavingsMsg'
Learn More

4-(Chloromethyl)-N-(2-fluorophenyl)benzamide, 97%, Thermo Scientific™

Catalog Number p-7039097
Click to view available options
Quantity:
100 mg
500 mg
This item is not returnable. View return policy
1094268-50-4
C14H11ClFNO
263.696
MFCD11544473
JIEHWZYCRNGLFE-UHFFFAOYSA-N
4-chloromethyl-n-2-fluorophenyl benzamide
43156289
4-(chloromethyl)-N-(2-fluorophenyl)benzamide
C1=CC=C(C(=C1)NC(=O)C2=CC=C(C=C2)CCl)F
This item is not returnable. View return policy
1094268-50-4
C14H11ClFNO
263.696
MFCD11544473
JIEHWZYCRNGLFE-UHFFFAOYSA-N
4-chloromethyl-n-2-fluorophenyl benzamide
43156289
4-(chloromethyl)-N-(2-fluorophenyl)benzamide
C1=CC=C(C(=C1)NC(=O)C2=CC=C(C=C2)CCl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store away from oxidizing agents. Store in a dry, cool conditions in a well sealed containers.
TRUSTED_SUSTAINABILITY
Quantity 100 mg
Formula Weight 263.69
Percent Purity 97%
Chemical Name or Material 4-(Chloromethyl)-N-(2-fluorophenyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'