Learn More
Alfa Aesar™ 4-Chloro-N-(2-methylbenzyl)-3-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65718.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15429856
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Chemical Identifiers
MFCD06199281
Specifications
4-Chloro-N-(2-methylbenzyl)-3-sulfamoylbenzamide
MFCD06199281
Solid
1g
338.81
95%