missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-3-(trifluoromethoxy)benzoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Chloro-3-(trifluoromethoxy)benzoic acid is used as pharmaceuticals intermediates.
Solubility
Sparingly Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4-Chloro-3-(trifluoromethoxy)benzoic acid is used as pharmaceuticals intermediates.
Solubility
Sparingly Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 123°C to 126°C |
Quantity | 1 g |
Solubility Information | Sparingly Soluble in water. |
Formula Weight | 240.57 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloro-3-(trifluoromethoxy)benzoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'