Learn More
4-chloro-3-nitrophenyl isothiocyanate, Thermo Scientific™
Catalog Number p-4391260
Click to view available options
Quantity:
5 g
10 g
25 g
127142-66-9
C7H3ClN2O2S
214.623
ZXGZBHIDSJXKLE-UHFFFAOYSA-N
4-chloro-3-nitrophenyl isothiocyanate, 4-chloro-3-nitrophenylisothiocyanate, acmc-1c0qb, 4-chloro-3-nitrobenzenisothiocyanate, #, benzene,1-chloro-4-isothiocyanato-2-nitro, 1-chloranyl-4-isothiocyanato-2-nitro-benzene
145581
1-chloro-4-isothiocyanato-2-nitrobenzene
C1=CC(=C(C=C1N=C=S)[N+](=O)[O-])Cl
127142-66-9
C7H3ClN2O2S
214.623
ZXGZBHIDSJXKLE-UHFFFAOYSA-N
4-chloro-3-nitrophenyl isothiocyanate, 4-chloro-3-nitrophenylisothiocyanate, acmc-1c0qb, 4-chloro-3-nitrobenzenisothiocyanate, #, benzene,1-chloro-4-isothiocyanato-2-nitro, 1-chloranyl-4-isothiocyanato-2-nitro-benzene
145581
1-chloro-4-isothiocyanato-2-nitrobenzene
C1=CC(=C(C=C1N=C=S)[N+](=O)[O-])Cl
Assay Percent Range 97%
Quantity 5 g
Formula Weight 214.63
Percent Purity 97%
Chemical Name or Material 4-Chloro-3-nitrophenyl isothiocyanate