Learn More
4-Chloro-3-methylbenzylamine hydrochloride, 97%, Thermo Scientific™
Catalog Number p-4229143
Click to view available options
Quantity:
1 g
5 g
1264198-68-6
C8H9N·ClH
192.09
WVNIGUUQWGGELM-UHFFFAOYSA-N
4-chloro-3-methylbenzylamine hydrochloride, 4-chloro-3-methylphenyl methanamine hydrochloride, 4-chloro-3-methyl-benzenemethanamine hydrochloride
110210981
(4-chloro-3-methylphenyl)methanamine;hydrochloride
CC1=C(C=CC(=C1)CN)Cl.Cl
1264198-68-6
C8H9N·ClH
192.09
WVNIGUUQWGGELM-UHFFFAOYSA-N
4-chloro-3-methylbenzylamine hydrochloride, 4-chloro-3-methylphenyl methanamine hydrochloride, 4-chloro-3-methyl-benzenemethanamine hydrochloride
110210981
(4-chloro-3-methylphenyl)methanamine;hydrochloride
CC1=C(C=CC(=C1)CN)Cl.Cl
Melting Point 252.3°C to 253.5°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 192.09
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Chloro-3-methylbenzylamine hydrochloride
compliance-icons
Product Identifier
  • 4-Chloro-3-methylbenzylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.