missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-3-fluorotoluene, 98%, Thermo Scientific™

Catalog Number p-7031261
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
5527-94-6
C7H6ClF
144.573
MFCD00143295
MHXNHUSVBYUTJL-UHFFFAOYSA-N
4-chloro-3-fluorotoluene, 3-fluoro-4-chlorotoluene, 2-fluoro-4-methylchlorobenzene, 1-chloro-2-fluoro-4-methyl-benzene, pubchem1611, 3-fluoro-4-chloro toluene, ksc493o2l, benzene, 1-chloro-2-fluoro-4-methyl
2736590
1-chloro-2-fluoro-4-methylbenzene
CC1=CC(=C(C=C1)Cl)F
This item is not returnable. View return policy
5527-94-6
C7H6ClF
144.573
MFCD00143295
MHXNHUSVBYUTJL-UHFFFAOYSA-N
4-chloro-3-fluorotoluene, 3-fluoro-4-chlorotoluene, 2-fluoro-4-methylchlorobenzene, 1-chloro-2-fluoro-4-methyl-benzene, pubchem1611, 3-fluoro-4-chloro toluene, ksc493o2l, benzene, 1-chloro-2-fluoro-4-methyl
2736590
1-chloro-2-fluoro-4-methylbenzene
CC1=CC(=C(C=C1)Cl)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN1993
Formula Weight 144.58
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-fluorotoluene
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'