missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Chloro-2-fluorophenylacetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Chloro-2-fluorophenylacetic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4-Chloro-2-fluorophenylacetic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Quantity | 1 g |
Solubility Information | Slightly soluble in water. |
Formula Weight | 188.59 |
Percent Purity | 97% |
Chemical Name or Material | 4-Chloro-2-fluorophenylacetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'