Learn More
Alfa Aesar™ 4-Chloro-2-fluoro-N-(4-iodophenyl)-5-sulfamoylbenzamide, 95%
Brand:  Alfa Aesar™ H65790.03
Additional Details : Weight : 0.00100kg
Quantity 1g
 View more versions of this product
Product Code. 15429906
  • 2830.00 NOK / 1g
Estimated Shipment date
from Supplier 19-08-2019
Specifications
4-Chloro-2-fluoro-N-(4-iodophenyl)-5-sulfamoylbenzamide
1g
Solid
Light Sensitive
454.64
95%