missing translation for 'onlineSavingsMsg'
Learn More

4-Chloro-1-butanol, tech. 85%, Thermo Scientific Chemicals

Catalog Number p-7044013
Click to view available options
Quantity:
50 g
250 g
928-51-8
C4H9ClO
108.57
MFCD00002967
HXHGULXINZUGJX-UHFFFAOYSA-N
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
13569
CHEBI:87310
4-chlorobutan-1-ol
OCCCCCl
928-51-8
C4H9ClO
108.57
MFCD00002967
HXHGULXINZUGJX-UHFFFAOYSA-N
4-chloro-1-butanol, 4-chlorobutanol, 1-butanol, 4-chloro, tetramethylene chlorohydrin, 4-chloro-1-butane-ol, 4-chlorbutan-1-ol, unii-42ei3i5ay0, 4-chlorbutan-1-ol german, ccris 7507, 4-chloro-butan-1-ol
13569
CHEBI:87310
4-chlorobutan-1-ol
OCCCCCl

4-Chloro-1-butanol is employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-MS technique.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chloro-1-butanol is employed as an internal standard in the analysis of genotoxic impurities in active pharmaceutical ingredients by GC-MS technique.

Solubility
Soluble in methanol, chloroform, and dimethyl sufoxide.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.089
Boiling Point 50°C to 52°C (1 mmHg)
Flash Point 36°C (97°F)
Linear Formula Cl(CH2)4OH
Refractive Index 1.457
Quantity 50 g
Beilstein 1731408
Solubility Information Soluble in methanol,chloroform,and dimethyl sufoxide.
Formula Weight 108.57
Percent Purity 85%
Grade Technical
Chemical Name or Material 4-Chloro-1-butanol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'