missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromopyridine hydrochloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in DMSO, Methanol and Water.
Notes
Store at -20°C. Hygroscopic. Incompatible materials are moisture, strong bases, strong oxidizing agents. Protect from humidity and air.
4-Bromopyridine hydrochloride is used as a useful synthetic intermediate. It is also used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
Soluble in DMSO, Methanol and Water.
Notes
Store at -20°C. Hygroscopic. Incompatible materials are moisture, strong bases, strong oxidizing agents. Protect from humidity and air.
Specifications
Melting Point | ∽270°C (decomposition) |
Quantity | 1 g |
Beilstein | 3621847 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in DMSO,Methanol and Water. |
Formula Weight | 194.46 |
Percent Purity | 99% |
Chemical Name or Material | 4-Bromopyridine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'