missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzyl bromide, 98+%, Thermo Scientific Chemicals
336.01 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 g
100 g
Description
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.
Solubility
Easily soluble in hot alcohol, ether, carbon disulfide, benzene and glacial acetic acid. Soluble in water and alcohol.
Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at 4°C. Incompatible with strong oxidizing agents. It is sensitive to light.
4-Bromobenzyl bromide is used to prepare a 20-member aminomethyl-substituted biaryl library via sequential N-alkylation of various amines.
Solubility
Easily soluble in hot alcohol, ether, carbon disulfide, benzene and glacial acetic acid. Soluble in water and alcohol.
Notes
Keep containers tightly closed in a dry, cool and well-ventilated place. Store at 4°C. Incompatible with strong oxidizing agents. It is sensitive to light.
Specifications
Melting Point | 60°C to 64°C |
Boiling Point | 115°C to 124°C (12 mmHg) |
Quantity | 25 g |
UN Number | UN1759 |
Beilstein | 606498 |
Merck Index | 14,1409 |
Solubility Information | Easily soluble in hot alcohol,ether,carbon disulfide,benzene and glacial acetic acid. Soluble in water and alcohol. |
Formula Weight | 249.94 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Bromobenzyl bromide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'