missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzamide, 97%, Thermo Scientific™

Catalog Number p-73065
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
698-67-9
C7H6BrNO
200.03
MFCD00007991
ZRWNRAJCPNLYAK-UHFFFAOYSA-N
p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
69683
4-bromobenzamide
C1=CC(=CC=C1C(=O)N)Br
This item is not returnable. View return policy
698-67-9
C7H6BrNO
200.03
MFCD00007991
ZRWNRAJCPNLYAK-UHFFFAOYSA-N
p-bromobenzamide, benzamide, 4-bromo, benzamide, p-bromo, p-bromobenzoic acid amide, 4-bromo-benzamide, 4-bromo benzamide, pubchem3737, acmc-209obe, 4-bromobenzamide, 4-bromobenzoic acid amide
69683
4-bromobenzamide
C1=CC(=CC=C1C(=O)N)Br
CAS Min % 96.0
CAS Max % 100.0
Melting Point 189°C to 194°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula BrC6H4CONH2
Quantity 10 g
Beilstein 09, 353
Formula Weight 200.03
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Bromobenzamide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Bromobenzamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'