missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde, 98+%, Thermo Scientific Chemicals

Catalog Number p-7021766
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1122-91-4
C7H5BrO
185.02
MFCD00003377
ZRYZBQLXDKPBDU-UHFFFAOYSA-N
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
70741
4-bromobenzaldehyde
C1=CC(=CC=C1C=O)Br
This item is not returnable. View return policy
1122-91-4
C7H5BrO
185.02
MFCD00003377
ZRYZBQLXDKPBDU-UHFFFAOYSA-N
p-bromobenzaldehyde, benzaldehyde, 4-bromo, benzaldehyde, p-bromo, 4-bromo-benzaldehyde, 4-bromo benzaldehyde, 4-bromobenzylaldehyde, p-bromo benzaldehyde, unii-4l8vm24f65, 4-brombenzaldehyde, 4-brombenzaldehyd
70741
4-bromobenzaldehyde
C1=CC(=CC=C1C=O)Br

4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzaldehyde is widely utilized as an intermediate for the preparation of agrochemicals, pharmaceuticals and other organic compounds. It is used as a precursor and reacts with sulfamethoxazole to prepare Schiff’s base, 4-[(4-Bromo-benzylidene)-amino]-N-(5-methyl-isoxazol-3-yl)-benzene sulfonamide, which can be used for the photo stability of polyvinyl chloride (PVC). It plays an important role in palladium-catalyzed mono- and bis-arylation of 3,4-(ethylenedioxy)thiophenes. It is employed in a cross-coupling study with potassium vinyltrifluoroborate.

Solubility
Soluble in ethanol and benzene. Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY
Melting Point 55°C to 58°C
Boiling Point 66°C to 68°C (2 mmHg)
Flash Point 108°C (226°F)
Odor Characteristic
Quantity 25 g
Beilstein 507100
Sensitivity Air Sensitive
Solubility Information Soluble in ethanol and benzene. Insoluble in water.
Formula Weight 185.03
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzaldehyde
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'