Learn More
Alfa Aesar™ 4-Bromo-3-fluoroanisole, 97%
Brand: Alfa Aesar™ H27319.06
Additional Details : CAS Number : 458-50-4 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
458-50-4 | |
205.026 | |
XANVIFOBBVAKCY-UHFFFAOYSA-N | |
3718444 | |
COC1=CC(=C(C=C1)Br)F |
C7H6BrFO | |
MFCD01310983 | |
4-bromo-3-fluoroanisole, 3-fluoro-4-bromo anisole, 3-fluoro-4-bromoanisol, 3-fluoro-4-bromo anisol, 1-bromo-2-fluoro-4-methoxy-benzene, benzene, 1-bromo-2-fluoro-4-methoxy, pharmabridge p-1018, rarechem al mz 0889, zlchem 365, pubchem1970 | |
1-bromo-2-fluoro-4-methoxybenzene |
Specifications
4-Bromo-3-fluoroanisole | |
217°C to 219°C | |
458-50-4 | |
MFCD01310983 | |
XANVIFOBBVAKCY-UHFFFAOYSA-N | |
1-bromo-2-fluoro-4-methoxybenzene | |
3718444 | |
97% |
85°C (185°F) | |
5g | |
C7H6BrFO | |
4-bromo-3-fluoroanisole, 3-fluoro-4-bromo anisole, 3-fluoro-4-bromoanisol, 3-fluoro-4-bromo anisol, 1-bromo-2-fluoro-4-methoxy-benzene, benzene, 1-bromo-2-fluoro-4-methoxy, pharmabridge p-1018, rarechem al mz 0889, zlchem 365, pubchem1970 | |
COC1=CC(=C(C=C1)Br)F | |
205.026 | |
205.03 |