Learn More
Alfa Aesar™ 4-Bromo-2-fluoro-1-nitrobenzene, 98%
Brand:  Alfa Aesar™ H61807.ME
Additional Details : CAS Number : 321-23-3 Weight : 0.00050kg
Quantity 500mg
 View more versions of this product
Product Code. 15448388
  • 212.00 NOK / 500mg
Estimated Shipment
22-12-2020
Chemical Identifiers
321-23-3
219.997
VQCWSOYHHXXWSP-UHFFFAOYSA-N
2783362
C1=CC(=C(C=C1Br)F)[N+](=O)[O-]
C6H3BrFNO2
MFCD01930221
4-bromo-2-fluoronitrobenzene, 2-fluoro-4-bromonitrobenzene, 4-bromo-2-fluoro-1-nitro-benzene, benzene, 4-bromo-2-fluoro-1-nitro, 1-bromo-3-fluoro-4-nitrobenzene, 2-fluoro-4-bromo-nitrobenzene, 4-bromo-2-fluoro-nitrobenzene, 4-bromo-2-fluornitrobenzole, 3-fluoro-4-nitrobromobenzene, 5-bromo-2-nitrofluorobenzene
4-bromo-2-fluoro-1-nitrobenzene
Specifications
4-Bromo-2-fluoro-1-nitrobenzene
84°C to 88°C
321-23-3
MFCD01930221
VQCWSOYHHXXWSP-UHFFFAOYSA-N
4-bromo-2-fluoro-1-nitrobenzene
2783362
Solid
Orange to Yellow
500mg
C6H3BrFNO2
4-bromo-2-fluoronitrobenzene, 2-fluoro-4-bromonitrobenzene, 4-bromo-2-fluoro-1-nitro-benzene, benzene, 4-bromo-2-fluoro-1-nitro, 1-bromo-3-fluoro-4-nitrobenzene, 2-fluoro-4-bromo-nitrobenzene, 4-bromo-2-fluoro-nitrobenzene, 4-bromo-2-fluornitrobenzole, 3-fluoro-4-nitrobromobenzene, 5-bromo-2-nitrofluorobenzene
C1=CC(=C(C=C1Br)F)[N+](=O)[O-]
219.997
220
98%