Learn More
4-Bromo-2-chlorobenzoic acid, 98%, Thermo Scientific™
Catalog Number p-7077298
Click to view available options
Quantity:
1 g
5 g
25 g
59748-90-2
C7H4BrClO2
235.47
JAVZWSOFJKYSDY-UHFFFAOYSA-N
benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d
108829
4-bromo-2-chlorobenzoic acid
C1=CC(=C(C=C1Br)Cl)C(=O)O
59748-90-2
C7H4BrClO2
235.47
JAVZWSOFJKYSDY-UHFFFAOYSA-N
benzoic acid, 4-bromo-2-chloro, 4-bromo-2-chlorobenzoicacid, 2-chloro-4-bromobenzoic acid, 4-bromo-2-chloro-benzoic acid, rarechem al bo 2371, attercop-chm at111793, zlchem 410, pubchem3586, acmc-1b03i, ksc274c2d
108829
4-bromo-2-chlorobenzoic acid
C1=CC(=C(C=C1Br)Cl)C(=O)O
Melting Point 171.0°C to 175.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 235.47
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chlorobenzoic acid
compliance-icons compliance-icons
Product Identifier
  • 4-Bromo-2-chlorobenzoic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.