Learn More
4-Bromo-2,6-dinitrotoluene, 97%, Thermo Scientific™
Catalog Number p-4813071
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
95192-64-6
C7H5BrN2O4
261.03
UOGCLPDKGPPDHM-UHFFFAOYSA-N
4-bromo-2,6-dinitrotoluene, 5-bromo-1,3-dinitro-2-methylbenzene, 5-bromo-2-methyl-1,3-dinitro-benzene, benzene, 5-bromo-2-methyl-1,3-dinitro, 2,6-dinitro-4-bromo-tolurene, 5-bromanyl-2-methyl-1,3-dinitro-benzene
13443186
5-bromo-2-methyl-1,3-dinitrobenzene
CC1=C(C=C(C=C1[N+](=O)[O-])Br)[N+](=O)[O-]
95192-64-6
C7H5BrN2O4
261.03
UOGCLPDKGPPDHM-UHFFFAOYSA-N
4-bromo-2,6-dinitrotoluene, 5-bromo-1,3-dinitro-2-methylbenzene, 5-bromo-2-methyl-1,3-dinitro-benzene, benzene, 5-bromo-2-methyl-1,3-dinitro, 2,6-dinitro-4-bromo-tolurene, 5-bromanyl-2-methyl-1,3-dinitro-benzene
13443186
5-bromo-2-methyl-1,3-dinitrobenzene
CC1=C(C=C(C=C1[N+](=O)[O-])Br)[N+](=O)[O-]
Melting Point 86°C to 88°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 261.03
Percent Purity 97%
Chemical Name or Material 4-Bromo-2, 6-dinitrotoluene
compliance-icons
Product Identifier
  • 4-Bromo-2,6-dinitrotoluene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.