missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1-indanone, 97%, Thermo Scientific Chemicals

Catalog Number p-4089816
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
15115-60-3
C9H7BrO
211.06
MFCD01719772
UVVYFYLSZIMKMC-UHFFFAOYSA-N
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
98713
4-bromo-2,3-dihydroinden-1-one
BrC1=CC=CC2=C1CCC2=O
This item is not returnable. View return policy
15115-60-3
C9H7BrO
211.06
MFCD01719772
UVVYFYLSZIMKMC-UHFFFAOYSA-N
4-bromo-1-indanone, 4-bromo-indan-1-one, 4-bromoindan-1-one, 4-bromo-2,3-dihydro-1h-inden-1-one, 1-indanone, 4-bromo, 4-bromohydrindone-1, 1h-inden-1-one, 4-bromo-2,3-dihydro, 4-bromoindanone, 4-bromo indanone, 4-bromo-indanone
98713
4-bromo-2,3-dihydroinden-1-one
BrC1=CC=CC2=C1CCC2=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 95°C to 99°C
Color Orange to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 211.05
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-1-indanone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'