missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-1,2-(methylenedioxy)benzene, 98%, Thermo Scientific Chemicals

Catalog Number p-7023894
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy
2635-13-4
C7H5BrO2
201.02
MFCD00005821
FBOYMIDCHINJKC-UHFFFAOYSA-N
4-bromo-1,2-methylenedioxy benzene, 5-bromobenzo d 1,3 dioxole, 4-bromo-1,2-methylenedioxybenzene, 1-bromo-3,4-methylenedioxy benzene, 3,4-methylenedioxybromobenzene, 5-bromo-2h-1,3-benzodioxole, 5-bromobenzo-1,3-dioxole, 5-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo, 3,4-methylenedioxy bromobenzene
75831
BrC1=CC=C2OCOC2=C1
This item is not returnable. View return policy
2635-13-4
C7H5BrO2
201.02
MFCD00005821
FBOYMIDCHINJKC-UHFFFAOYSA-N
4-bromo-1,2-methylenedioxy benzene, 5-bromobenzo d 1,3 dioxole, 4-bromo-1,2-methylenedioxybenzene, 1-bromo-3,4-methylenedioxy benzene, 3,4-methylenedioxybromobenzene, 5-bromo-2h-1,3-benzodioxole, 5-bromobenzo-1,3-dioxole, 5-bromo-benzo 1,3 dioxole, 1,3-benzodioxole, 5-bromo, 3,4-methylenedioxy bromobenzene
75831
BrC1=CC=C2OCOC2=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.669
Boiling Point 228°C
Flash Point >110°C (230°F)
Refractive Index 1.584
Quantity 5 g
Beilstein 127407
IUPAC Name 5-bromo-2H-1,3-benzodioxole
Formula Weight 201.02
Percent Purity 98%
Chemical Name or Material 4-Bromo-1,2-(methylenedioxy)benzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'