missing translation for 'onlineSavingsMsg'
Learn More

4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3735607
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5728-52-9
MFCD00004351
QRZAKQDHEVVFRX-UHFFFAOYSA-N
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
3332
CHEBI:31597
2-(4-phenylphenyl)acetic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O
This item is not returnable. View return policy
5728-52-9
MFCD00004351
QRZAKQDHEVVFRX-UHFFFAOYSA-N
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
3332
CHEBI:31597
2-(4-phenylphenyl)acetic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 156°C to 160°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C14H12O2
Linear Formula C6H5C6H4CH2CO2H
Quantity 10 g
Beilstein 09, I, 284
Merck Index 15, 3985
Molecular Weight (g/mol) 212.25
Formula Weight 212.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 4-Biphenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Biphenylacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'