missing translation for 'onlineSavingsMsg'
Learn More

4-Benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0-1.5 mmol/g on Merrifield resin, Thermo Scientific™

Catalog Number p-7047788
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
679428-14-9
C16H16O4
272.30
MFCD02683435
YHMXRPWCVSRREV-UHFFFAOYSA-N
4-benzyloxy-2,6-dimethoxybenzaldehyde, 4-benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0-1.5 mmol/g on merrifield resin, fdmp resin, syringaldehyde, polymer-bound, stratospheres pl-fdmp resin, 2,6-dimethoxy-4-phenylmethoxy-benzaldehyde, 4-formyl-3,5-dimethoxyphenoxymethyl polystyrene, benzaldehyde, 2,6-dimethoxy-4-phenylmethoxy, 4-hydroxy-2,6-dimethoxybenzaldehyde, polymer-bound
4607337
2,6-dimethoxy-4-phenylmethoxybenzaldehyde
COC1=CC(OCC2=CC=CC=C2)=CC(OC)=C1C=O
This item is not returnable. View return policy
679428-14-9
C16H16O4
272.30
MFCD02683435
YHMXRPWCVSRREV-UHFFFAOYSA-N
4-benzyloxy-2,6-dimethoxybenzaldehyde, 4-benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0-1.5 mmol/g on merrifield resin, fdmp resin, syringaldehyde, polymer-bound, stratospheres pl-fdmp resin, 2,6-dimethoxy-4-phenylmethoxy-benzaldehyde, 4-formyl-3,5-dimethoxyphenoxymethyl polystyrene, benzaldehyde, 2,6-dimethoxy-4-phenylmethoxy, 4-hydroxy-2,6-dimethoxybenzaldehyde, polymer-bound
4607337
2,6-dimethoxy-4-phenylmethoxybenzaldehyde
COC1=CC(OCC2=CC=CC=C2)=CC(OC)=C1C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Sensitivity Air Sensitive
Chemical Name or Material 4-Benzyloxy-2,6-dimethoxybenzaldehyde, polymer-supported, 1.0 to 1.5mmol/g on Merrifield resin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'