Learn More
(4-Aminophenylthio)acetic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It can react with 2-phenyl-benzo[d][1,3]oxazin-4-one to get 2-(Phenyl-4-(3H)-quinazolyl)-phenyl-4′-mercaptoacetic acid. This reaction needs solvents pyridine and H2O by heating.
Solubility
Soluble in methanol, acetone, toluene, n-octanol, acetonitrile, ethyl acetate, dichloromethane.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
It can react with 2-phenyl-benzo[d][1,3]oxazin-4-one to get 2-(Phenyl-4-(3H)-quinazolyl)-phenyl-4′-mercaptoacetic acid. This reaction needs solvents pyridine and H2O by heating.
Solubility
Soluble in methanol, acetone, toluene, n-octanol, acetonitrile, ethyl acetate, dichloromethane.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | ∼200°C (decomposition) |
Quantity | 1 g |
Beilstein | 2720454 |
Solubility Information | Soluble in methanol,acetone,toluene,n-octanol,acetonitrile,ethyl acetate,dichloromethane. |
Formula Weight | 183.23 |
Percent Purity | 97% |
Chemical Name or Material | (4-Aminophenylthio)acetic acid |