missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-3-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7024488
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
1588-83-6
C7H6N2O4
182.135
MFCD00017009
ZZNAYFWAXZJITH-UHFFFAOYSA-N
4-amino-3-nitrobenzoic acid, 3-nitro-4-aminobenzoic acid, benzoic acid, 4-amino-3-nitro, 4-carboxy-2-nitroaniline, 3-nitro-4-aminobenozic acid, 4-amino-3-nitro-benzoic acid, 4-amino-3-nitrobenzoicacid, pubchem4982, rarechem al bo 0261, 2-nitro-4-carboxyaniline
74115
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])N
This item is not returnable. View return policy
1588-83-6
C7H6N2O4
182.135
MFCD00017009
ZZNAYFWAXZJITH-UHFFFAOYSA-N
4-amino-3-nitrobenzoic acid, 3-nitro-4-aminobenzoic acid, benzoic acid, 4-amino-3-nitro, 4-carboxy-2-nitroaniline, 3-nitro-4-aminobenozic acid, 4-amino-3-nitro-benzoic acid, 4-amino-3-nitrobenzoicacid, pubchem4982, rarechem al bo 0261, 2-nitro-4-carboxyaniline
74115
C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼290°C (decomposition)
Quantity 10 g
Beilstein 645206
Formula Weight 182.14
Percent Purity 97%
Chemical Name or Material 4-Amino-3-nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'