Learn More
4-Amino-1H-pyrazolo[3,4-d]pyrimidine, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Amino-1H-pyrazolo[3,4-d]pyrimidine decreases serum cholesterol markedly in rats.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
4-Amino-1H-pyrazolo[3,4-d]pyrimidine decreases serum cholesterol markedly in rats.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | >310°C (decomposition) |
Odor | Odorless |
Quantity | 250 mg |
UN Number | UN2811 |
Beilstein | 5824 |
Solubility Information | Insoluble in water. |
Formula Weight | 135.13 |
Percent Purity | 98% |
Chemical Name or Material | 4-Amino-1H-pyrazolo[3,4-d]pyrimidine |