missing translation for 'onlineSavingsMsg'
Learn More

4-Amino-1-Boc-piperidine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-7047812
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
183673-71-4
C11H20N2O4
244.29
MFCD01318728
YNHLVALLAURVJF-UHFFFAOYSA-N
1-boc-4-aminopiperidine-4-carboxylic acid, 4-amino-1-boc-piperidine-4-carboxylic acid, 4-amino-1-boc-isonipecotic acid, 4-amino-1-tert-butoxycarbonyl piperidine-4-carboxylic acid, h-pip boc-oh, 4,4-acp 1-n-boc, 4-amino-piperidine-1,4-dicarboxylic acid mono-tert-butyl ester, 1-boc-4-amino-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxycarbonyl-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxy carbonyl piperidine-4-carboxylic acid
693797
CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O
This item is not returnable. View return policy
183673-71-4
C11H20N2O4
244.29
MFCD01318728
YNHLVALLAURVJF-UHFFFAOYSA-N
1-boc-4-aminopiperidine-4-carboxylic acid, 4-amino-1-boc-piperidine-4-carboxylic acid, 4-amino-1-boc-isonipecotic acid, 4-amino-1-tert-butoxycarbonyl piperidine-4-carboxylic acid, h-pip boc-oh, 4,4-acp 1-n-boc, 4-amino-piperidine-1,4-dicarboxylic acid mono-tert-butyl ester, 1-boc-4-amino-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxycarbonyl-4-piperidinecarboxylic acid, 4-amino-1-tert-butoxy carbonyl piperidine-4-carboxylic acid
693797
CC(C)(C)OC(=O)N1CCC(N)(CC1)C(O)=O

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY
Melting Point ∽290°C (decomposition)
Quantity 250 mg
Beilstein 7588844
IUPAC Name 4-amino-1-[(tert-butoxy)carbonyl]piperidine-4-carboxylic acid
Formula Weight 244.29
Percent Purity 98%
Chemical Name or Material 4-Amino-1-Boc-piperidine-4-carboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'