Learn More
4'-Acetamido-3'-bromoacetophenone, 98%, Thermo Scientific™
Catalog Number p-7044658
Click to view available options
Quantity:
5 g
25 g
101209-08-9
C10H10BrNO2
256.10
MFCD00051781
PMYJAVHDFDKJBS-UHFFFAOYSA-N
4-acetamido-3-bromoacetophenone, 4'-acetamido-3'-bromoacetophenone, n-4-acetyl-2-bromophenyl acetamide, 4-acetyl-2-bromoacetanilide, 4'-acetamido-3-bromoacetophenone, 3'-bromo-4'-acetaminoacetophenone, n-2-bromo-4-acetylphenyl acetamide, n-4-acetyl-2-bromo-phenyl acetamide, n-4-acetyl-2-bromo-phenyl-acetamide
3682474
N-(4-acetyl-2-bromophenyl)acetamide
CC(=O)NC1=C(Br)C=C(C=C1)C(C)=O
101209-08-9
C10H10BrNO2
256.10
MFCD00051781
PMYJAVHDFDKJBS-UHFFFAOYSA-N
4-acetamido-3-bromoacetophenone, 4'-acetamido-3'-bromoacetophenone, n-4-acetyl-2-bromophenyl acetamide, 4-acetyl-2-bromoacetanilide, 4'-acetamido-3-bromoacetophenone, 3'-bromo-4'-acetaminoacetophenone, n-2-bromo-4-acetylphenyl acetamide, n-4-acetyl-2-bromo-phenyl acetamide, n-4-acetyl-2-bromo-phenyl-acetamide
3682474
N-(4-acetyl-2-bromophenyl)acetamide
CC(=O)NC1=C(Br)C=C(C=C1)C(C)=O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Melting Point 139°C to 141°C
Quantity 5 g
Beilstein 3272108
Formula Weight 256.1
Percent Purity 98%
Chemical Name or Material 4'-Acetamido-3'-bromoacetophenone