missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethylpyrimidine, 94%, Thermo Scientific Chemicals

Catalog Number p-7080672
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1558-17-4
C6H8N2
108.14
MFCD00006116
LSBIUXKNVUBKRI-UHFFFAOYSA-N
4,6-dimethyl-pyrimidine, pyrimidine, 4,6-dimethyl, unii-95n99d3vhb, pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci, dimethylpyrimidinetech, zlchem 778, pubchem21498, 4,6-dimethyl-pyrimidin, 4,6-dimethyl-pyrmidine, acmc-1bwo9
15257
4,6-dimethylpyrimidine
CC1=CC(C)=NC=N1
This item is not returnable. View return policy
1558-17-4
C6H8N2
108.14
MFCD00006116
LSBIUXKNVUBKRI-UHFFFAOYSA-N
4,6-dimethyl-pyrimidine, pyrimidine, 4,6-dimethyl, unii-95n99d3vhb, pyrimidine, 4,6-dimethyl-6ci,7ci,8ci,9ci, dimethylpyrimidinetech, zlchem 778, pubchem21498, 4,6-dimethyl-pyrimidin, 4,6-dimethyl-pyrmidine, acmc-1bwo9
15257
4,6-dimethylpyrimidine
CC1=CC(C)=NC=N1

4,6-Dimethylpyrimidine is used to produce (6-methyl-pyrimidin-4-yl)-pyruvic acid ethyl ester. It is used as pharmaceutical intermediate. It is also used in agrochemical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dimethylpyrimidine is used to produce (6-methyl-pyrimidin-4-yl)-pyruvic acid ethyl ester. It is used as pharmaceutical intermediate. It is also used in agrochemical and dyestuff field.

Solubility
Soluble in water at 20°C.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 0.98
Boiling Point 153°C to 155°C
Flash Point 46°C (114°F)
Odor Recognizable
Refractive Index 1.49
Quantity 1 g
UN Number UN1993
Beilstein 107068
Solubility Information Soluble in water at 20°C.
Formula Weight 108.14
Percent Purity 94%
Chemical Name or Material 4,6-Dimethylpyrimidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'