missing translation for 'onlineSavingsMsg'
Learn More

4,6-Dimethyldibenzothiophene, 97%, Thermo Scientific Chemicals

Catalog Number p-7036971
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
1207-12-1
C14H12S
212.31
MFCD00216264
KMPJENUWHPZRGZ-UHFFFAOYSA-N
dibenzothiophene, 4,6-dimethyl, 4,6-dimethyldibenzo b,d thiophene, 4,6-dmdbt, 6,10-dimethyl-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4,6-dimethyl-dibenzothiophen, ksc491i3n, bidd:gt0307, 4,6-dimethyl dibenzo b thiophene, 4,6-dimethyldibenzothiophene, 4,6-dimethylbenzo b benzo b thiophene
1268103
4,6-dimethyldibenzothiophene
CC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1
This item is not returnable. View return policy
1207-12-1
C14H12S
212.31
MFCD00216264
KMPJENUWHPZRGZ-UHFFFAOYSA-N
dibenzothiophene, 4,6-dimethyl, 4,6-dimethyldibenzo b,d thiophene, 4,6-dmdbt, 6,10-dimethyl-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4,6-dimethyl-dibenzothiophen, ksc491i3n, bidd:gt0307, 4,6-dimethyl dibenzo b thiophene, 4,6-dimethyldibenzothiophene, 4,6-dimethylbenzo b benzo b thiophene
1268103
4,6-dimethyldibenzothiophene
CC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1

4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 153°C to 157°C
Quantity 250 mg
Formula Weight 212.31
Percent Purity 97%
Chemical Name or Material 4,6-Dimethyldibenzothiophene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'