missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Isopropylidenediphenol, 97%, Thermo Scientific Chemicals

Catalog Number p-151491
Click to view available options
Quantity:
100 g
500 g
2 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy
80-05-7
C15H16O2
228.29
MFCD00002366
IISBACLAFKSPIT-UHFFFAOYSA-N
bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian
6623
CHEBI:33216
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
This item is not returnable. View return policy
80-05-7
C15H16O2
228.29
MFCD00002366
IISBACLAFKSPIT-UHFFFAOYSA-N
bisphenol a, 2,2-bis 4-hydroxyphenyl propane, 4,4'-isopropylidenediphenol, diphenylolpropane, bisphenol, diano, 4,4'-bisphenol a, 4,4'-propane-2,2-diyl diphenol, biphenol a, dian
6623
CHEBI:33216
4-[2-(4-hydroxyphenyl)propan-2-yl]phenol
CC(C)(C1=CC=C(O)C=C1)C1=CC=C(O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 155.0°C to 158.0°C
Color Yellow to Orange
Boiling Point 220.0°C (4.0 mmHg)
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Linear Formula (CH3)2C(C6H4OH)2
Quantity 100 g
Beilstein 06,1011
Merck Index 15,1298
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in dilute alkalies, soluble in alcohol, slightly soluble in ccl4
Formula Weight 228.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4,4′-Isopropylidenediphenol, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 4,4'-Isopropylidenediphenol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361f-Suspected of damaging fertility.
  • H411-Toxic to aquatic life with long lasting effects.
  • H335-May cause respiratory irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'