missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Fluorobenzyl)piperidine hydrochloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Reactant in the synthesis of 4-(Substituted-benzyl)piperidines and (±)-3-(Substituted-benzyl)pyrrolidines. It an important and valuable pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant in the synthesis of 4-(Substituted-benzyl)piperidines and (±)-3-(Substituted-benzyl)pyrrolidines. It an important and valuable pharmaceutical intermediate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Reactant in the synthesis of 4-(Substituted-benzyl)piperidines and (±)-3-(Substituted-benzyl)pyrrolidines. It an important and valuable pharmaceutical intermediate.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat.
Specifications
Quantity | 25 g |
IUPAC Name | 4-[(4-fluorophenyl)methyl]piperidine;hydrochloride |
Formula Weight | 229.72 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 4-(4-Fluorobenzyl)piperidine hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'