Learn More
4,4'-Dithio-bis(2,6-di-tert-butylphenol), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
4,4'-Dithio-bis(2,6-di-tert-butylphenol)
Specifications
Specifications
| Chemical Name or Material | 4,4'-Dithio-bis(2,6-di-tert-butylphenol) |
| CAS | 6386-58-9 |
| Analyte or Component Names | 4,4'-Dithio-bis(2,6-di-tert-butylphenol) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H42 O2 S2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Phenol, 4,4'-dithiobis[2,6-bis(1,1-dimethylethyl)-, Phenol, 4,4'-dithiobis[2,6-di-tert-butyl- (7CI,8CI), 4,4'-Dithiobis[2,6-di-tert-butylphenol], Bis(3,5-di-tert-butyl-4-hydroxybenzene) disulfide, Bis(3,5-di-tert-butyl-4-hydroxyphenyl) disulfide, Bis(3,5-di-tert-butylphen-4-ol) disulfide, Bis(4-hydroxy-3,5-di-tert-butylphenyl) disulfide, 4,4'-Dithio-bis(2,6-di-tert-butylphenol), Probucol Related Compound B |
| InChI Formula | InChI=1S/C28H42O2S2/c1-25(2,3)19-13-17(14-20(23(19)29)26(4,5)6)31-32-18-15-21(27(7,8)9)24(30)22(16-18)28(10,11)12/h13-16,29-30H,1-12H3 |
| SMILES | CC(C)(C)c1cc(SSc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C |
| IUPAC Name | 2,6-ditert-butyl-4-[(3,5-ditert-butyl-4-hydroxyphenyl)disulfanyl]phenol |
| Show More |