Learn More
Memmert™ 4-20Ma Current Loop Interface Temperature 0-260°C Of One Pt100 In Chamber, For U75
Memmert™ Temperature Output for external temperature monitoring.
Brand:  Memmert™ U75V6
Additional Details : Weight : 0.01100kg
Product Code. 16356557
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • 4 to 20 mA current loop interface
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring
Specifications
Specifications
Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Current Loop Interface
4 to 20 mA