missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Hydroxyethyl)pyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7058402
660.03 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5344-27-4
C7H9NO
123.155
MFCD00129038
DWPYQDGDWBKJQL-UHFFFAOYSA-N
4-pyridineethanol, 2-pyridin-4-yl ethanol, 4-2-hydroxyethyl pyridine, 4-ethanolpyridine, 2-4-pyridyl ethanol, 2-pyridin-4-yl ethan-1-ol, 2-pyridin-4-yl-ethanol, 2-gamma-pyridyl ethanol, unii-i89rdq5yd9
72921
2-pyridin-4-ylethanol
C1=CN=CC=C1CCO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
5344-27-4
C7H9NO
123.155
MFCD00129038
DWPYQDGDWBKJQL-UHFFFAOYSA-N
4-pyridineethanol, 2-pyridin-4-yl ethanol, 4-2-hydroxyethyl pyridine, 4-ethanolpyridine, 2-4-pyridyl ethanol, 2-pyridin-4-yl ethan-1-ol, 2-pyridin-4-yl-ethanol, 2-gamma-pyridyl ethanol, unii-i89rdq5yd9
72921
2-pyridin-4-ylethanol
C1=CN=CC=C1CCO

4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(2-Hydroxyethyl)pyridine acts as a reagent for the protection of carboxyl groups as their 2-(4-pyridyl)ethyl esters in organic synthesis. Further, it is used as an active pharmaceutical ingredient intermediate.

Solubility
Miscible with water.

Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents, bases and mineral acids.
TRUSTED_SUSTAINABILITY
Melting Point 8°C to 10°C
Density 1.093
Boiling Point 121°C to 122°C (2 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.538
Quantity 1 g
Beilstein 111361
Sensitivity Hygroscopic
Solubility Information Miscible with water.
Formula Weight 123.16
Percent Purity 98%
Chemical Name or Material 4-(2-Hydroxyethyl)pyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'